6-Bromonicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid
A169647-1G
A169647-100G
A169647-10G
A169647-500G
A169647-25G
A169647-5G
103806-394EA
6
USD
103806-394
103844-654
77081-742
77273-662
77273-664
77273-666
77300-984
77300-986
6-Bromonicotinic acid 98%
6-Bromonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Boiling Pt: 343.2 °C (760 mm Hg) Melting Pt: 200…203 °C Flash Pt: 161.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD01927100 CAS Number: 6311-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...