6-Bromonicotinamide 98%

Supplier: Ambeed
Warning

A205150-10G A205150-250MG A205150-1G A205150-5G
103817-604EA 21 USD
103817-604 103848-268 76975-330 76975-332 77086-468 77086-470
6-Bromonicotinamide 98%
6-Bromonicotinamide

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O
MW: 201.0200 g/mol
Boiling Pt: 352.4 °C (760 mm Hg)
Melting Pt: 238…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00266840
CAS Number: 889676-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR