2-Bromo-5-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde

A104469-100G A104469-10G A104469-25G A104469-500G A104469-1G A104469-5G
103796-714EA 9 USD
103796-714 103822-428 76994-732 76994-734 76994-736 76994-738 77283-212 77309-294
2-Bromo-5-formylpyridine 97%
2-Bromo-5-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Boiling Pt: 284.1 °C (760 mm Hg)
Melting Pt: 104…110 °C
Flash Pt: 125.6 °C
Storage Temperature: Ambient
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR