6-Bromo-2-naphthylamine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromonaphthalen-2-amine, 2-Amino-6-bromonaphthalene

A580048-25G A580048-5G A580048-100G A580048-10G A580048-250MG A580048-1G
103828-860EA 7 USD
103828-860 103840-008 76844-142 76844-144 77117-154 77117-156 77117-158 77307-868
6-Bromo-2-naphthylamine 98%
6-Bromo-2-naphthylamine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.0812 g/mol
Boiling Pt: 364.4 °C (760 mm Hg)
Flash Pt: 174.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01026466
CAS Number: 7499-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR