1-Amino-6-bromoisoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromoisoquinolin-1-ylamine

A323648-1G A323648-5G A323648-100MG A323648-250MG
103825-348EA 96 USD
103825-348 103836-290 76839-204 76839-206 77015-294 77015-296
1-Amino-6-bromoisoquinoline 97%
1-Amino-6-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0700 g/mol
Boiling Pt: 380.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07374395
CAS Number: 215453-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR