6-Bromoisoquinolin-1(2H)-one 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A422731-1G A422731-5G A422731-10G A422731-25G A422731-250MG
103827-424EA 9 USD
103827-424 103838-472 76842-098 76842-100 77016-476 77016-478 77684-181
6-Bromoisoquinolin-1(2H)-one 97%, Ambeed.Inc
6-Bromoisoquinolin-1(2H)-one

Precaución: For research use only.

Formula: C₉H₆BrNO
Boiling Pt: 442.6 °C (760 mm Hg)
Flash Pt: 221.498 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07374394
CAS Number: 82827-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR