6-Bromoisoindolin-1-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2,3-dihydro-1H-isoindol-1-one

A345051-1G A345051-25G A345051-5G A345051-100G A345051-10G A345051-250MG
103825-916EA 11 USD
103825-916 103836-898 76839-952 76839-954 76839-956 76978-448 76978-450 76978-452
6-Bromoisoindolin-1-one 97%
6-Bromoisoindolin-1-one

Precaución: For research use only

Formula: C₈H₆BrNO
MW: 212.0433 g/mol
Boiling Pt: 431 °C (760 mm Hg)
Flash Pt: 214.881 °C
Storage Temperature: Ambient
MDL Number: MFCD11848599
CAS Number: 675109-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR