6-Bromooxindole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

A146590-100G A146590-10G A146590-1G A146590-25G A146590-5G A146590-500G
103803-686EA 6 USD
103803-686 103841-780 77235-226 77235-228 77299-536 77317-112 77317-114 77746-030
6-Bromooxindole 98%
6-Bromooxindole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 217…221 °C
Flash Pt: 161.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179605
CAS Number: 99365-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR