6-Bromoindole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1H-indole

A186285-25G A186285-5G A186285-100G A186285-10G A186285-500G A186285-1G
103815-930EA 5 USD
103815-930 103846-508 77084-162 77084-164 77274-314 77274-316 77274-318 77302-028
6-Bromoindole 98%
6-Bromoindole

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 316.9 °C (760 mm Hg)
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR