6-Bromoimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A112997-10G A112997-250MG A112997-25G A112997-1G A112997-5G
103798-144EA 23 USD
103798-144 103823-978 76766-000 76766-002 76766-004 77310-812 77310-814
6-Bromoimidazo[1,2-a]pyrazine 97%
6-Bromoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD09750036
CAS Number: 912773-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR