6-Bromocaproic acid 97%
Supplier: Ambeed
|
Danger
|
A544674-25G
A544674-10G
A544674-5G
A544674-100G
103839-778EA
5
USD
103839-778
76843-842
77169-688
77307-786
77594-670
6-Bromocaproic acid 97%
6-Bromocaproic acid
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₄COOH Boiling Pt: 279.2 °C (760 mm Hg) Melting Pt: 32…34 °C Flash Pt: 95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004422 CAS Number: 4224-70-8 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...