6-Bromo-1-hexanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane
A153717-100G
A153717-10G
A153717-500G
A153717-1G
A153717-25G
A153717-5G
A153717-1000G
103804-556EA
5
USD
103804-556
103842-732
76973-230
76973-232
76973-234
77318-410
77318-412
77318-414
77549-948
6-Bromo-1-hexanol 97%
6-Bromo-1-hexanol
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₄CH₂OH Boiling Pt: 198.9 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002983 CAS Number: 4286-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...