6-Bromocinnoline 95%

Supplier: Ambeed
Warning

A143870-100MG A143870-1G
103803-334EA 306 USD
103803-334 76972-730 77316-642
6-Bromocinnoline 95%
6-Bromocinnoline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MW: 209.04 g/mol
Boiling Pt: 325 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09834644
CAS Number: 318276-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR