6-Bromochroman-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4-chromanone

A307269-1G A307269-25G A307269-5G A307269-100G A307269-250MG
103814-496EA 6 USD
103814-496 103835-674 76838-408 76838-410 76838-412 77263-130 77263-132
6-Bromochroman-4-one 97%
6-Bromochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.0547 g/mol
Boiling Pt: 336.6 °C (760 mm Hg)
Melting Pt: 77…83 °C
Flash Pt: 157.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00546803
CAS Number: 49660-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR