6-Bromobenzo[b]thiophene 98%
Supplier: Ambeed
|
Warning
|
A191295-100G
A191295-10G
A191295-250MG
A191295-1G
A191295-25G
A191295-5G
103816-362EA
10
USD
103816-362
103846-964
77029-802
77029-804
77029-806
77084-792
77084-794
77084-796
6-Bromobenzo[b]thiophene 98%
6-Bromobenzo[b]thiophene
Precaución: for Research Use Only
Formula:
C₈H₅BrS Boiling Pt: 284.7 °C (760 mm Hg) Melting Pt: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD11226847 CAS Number: 17347-32-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...