6-Bromobenzofuran 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1-benzofuran

A236465-250MG A236465-5G A236465-100MG
103811-096EA 52 USD
103811-096 103850-868 76692-180 77089-916 77593-518
6-Bromobenzofuran 97%
6-Bromobenzofuran

Precaución: for Research Use Only

Formula: C₈H₅BrO
MW: 197.029 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669477
CAS Number: 128851-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR