4-Bromo-1,8-naphthalic anhydride 96%

Supplier: Ambeed
Warning

Synonyms: 6-Bromobenzo[de]isochromene-1,3-dione

A202986-25G A202986-100G A202986-10G A202986-5G A202986-1000G
103848-084EA 5 USD
103848-084 77086-196 77287-718 77287-720 77302-842 77748-281
4-Bromo-1,8-naphthalic anhydride 96%
4-Bromo-1,8-naphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₅BrO₃
MW: 277.0703 g/mol
Melting Pt: 217…219 °C
Flash Pt: 236.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006927
CAS Number: 81-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR