6-Bromobenzo[d]thiazol-2(3H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-benzothiazolinone

A133094-100G A133094-10G A133094-1G A133094-25G A133094-5G
103801-824EA 8 USD
103801-824 76811-328 76811-330 77314-422 77314-424 77314-426
6-Bromobenzo[d]thiazol-2(3H)-one 95%
6-Bromobenzo[d]thiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
MW: 230.08 g/mol
Melting Pt: 229…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00239363
CAS Number: 62266-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR