6-Bromobenzo[d]oxazole 98%

Supplier: Ambeed
Warning

A143305-100MG A143305-250MG A143305-1G A143305-25G A143305-5G
103803-264EA 19 USD
103803-264 76972-696 76972-698 77316-520 77316-522 77316-524
6-Bromobenzo[d]oxazole 98%
6-Bromobenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₇H₄BrNO
Flash Pt: 105.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092891
CAS Number: 375369-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR