6-Bromo-2(3H)-benzoxazolone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-benzoxazolinone, 6-Bromo-2-benzoxazolone, 6-Bromo-2-hydroxybenzoxazole, 6-Bromobenzo[d]oxazol-2(3H)-one

A274337-100G A274337-10G A274337-1G A274337-25G A274337-5G
103813-192EA 10 USD
103813-192 103853-116 77001-756 77001-758 77092-940 77092-942 77092-944
6-Bromo-2(3H)-benzoxazolone 98%
6-Bromo-2(3H)-benzoxazolone

Precaución: for Research Use Only

Formula: C₇H₄BrNO₂
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00694727
CAS Number: 19932-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR