6-Bromobenzo[d]isoxazol-3-amine 97%

Supplier: Ambeed
Warning

A118446-100MG A118446-10G A118446-250MG A118446-5G A118446-1G
103799-062EA 26 USD
103799-062 76766-306 76766-308 76766-310 77297-604 77311-732
6-Bromobenzo[d]isoxazol-3-amine 97%
6-Bromobenzo[d]isoxazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.0314 g/mol
Flash Pt: 178.236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09788587
CAS Number: 177995-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR