6-Bromopiperonal 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromobenzo[d][1,3]dioxole-5-carbaldehyde
A735142-25G
A735142-5G
A735142-100G
A735142-10G
A735142-500G
A735142-1G
103840-810EA
14
USD
103840-810
76845-036
76845-038
77105-128
77105-130
77105-132
77308-234
6-Bromopiperonal 98%
6-Bromopiperonal
Precaución: for Research Use Only
Formula:
C₈H₅BrO₃ MW: 229.03 g/mol Boiling Pt: 313.9 °C (760 mm Hg) Melting Pt: 128…132 °C Flash Pt: 143.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00022952 CAS Number: 15930-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...