6-Bromo-8-chloroquinoline 97%

Supplier: Ambeed
Warning

A135650-1G A135650-5G
103802-192EA 14 USD
103802-192 77315-036 77315-038
6-Bromo-8-chloroquinoline 97%
6-Bromo-8-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Boiling Pt: 327.8±22.0 °C (760 mm Hg)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD12913961
CAS Number: 16567-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR