6-Bromo-8-chloroimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A440728-100MG A440728-10G A440728-250MG A440728-1G A440728-25G A440728-5G
103827-666EA 44 USD
103827-666 103838-732 76789-268 76789-270 76789-272 76842-496 76842-498 76842-500
6-Bromo-8-chloroimidazo[1,2-a]pyrazine 97%
6-Bromo-8-chloroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846485
CAS Number: 1208083-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR