6-Bromo-7-methylquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-7-methyl-1-azanaphthalene

A157848-10G A157848-250MG A157848-1G A157848-5G
103805-052EA 27 USD
103805-052 103843-258 76824-004 76824-006 77300-296 77300-298
6-Bromo-7-methylquinoline 97%
6-Bromo-7-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.081 g/mol
Boiling Pt: 312.7±22.0 °C (760 mm Hg)
Flash Pt: 142.9 °C
Storage Temperature: Ambient
MDL Number: MFCD18260428
CAS Number: 122759-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR