6-Bromo-7-methylimidazo[1,2-a]pyridine 95%
Supplier: Ambeed
|
Warning
|
A422408-1G
A422408-10G
A422408-250MG
103827-416EA
28
USD
103827-416
103838-464
76842-090
76877-142
76877-144
6-Bromo-7-methylimidazo[1,2-a]pyridine 95%
6-Bromo-7-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09258769 CAS Number: 116355-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...