6-Bromo-5-methylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A491418-1G A491418-5G A491418-10G A491418-250MG A491418-25G
103828-218EA 15 USD
103828-218 103839-290 76843-252 76843-254 76991-126 76991-128 77684-312
6-Bromo-5-methylpyridin-2-amine 95%
6-Bromo-5-methylpyridin-2-amine

Precaución: For research use only.

Formula: C₆H₇BrN₂
Boiling Pt: 287.3±35.0 °C (760 mm Hg)
Melting Pt: 96…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20704687
CAS Number: 89466-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR