6-Bromo-5-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

A252966-10G A252966-250MG A252966-1G A252966-25G A252966-5G
103812-068EA 26 USD
103812-068 103851-916 77031-584 77031-586 77091-340 77091-342 77091-344
6-Bromo-5-methyl-1H-indazole 98%
6-Bromo-5-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Boiling Pt: 344.6 °C (760 mm Hg)
Melting Pt: 176…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09026997
CAS Number: 1000343-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR