6-Bromo-5-methoxypyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-methoxypicolinic acid

A163884-100MG A163884-250MG A163884-1G A163884-10G
103805-782EA 61 USD
103805-782 103844-046 76768-734 76768-736 77320-170 77625-612
6-Bromo-5-methoxypyridine-2-carboxylic acid 95%
6-Bromo-5-methoxypyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.04 g/mol
Boiling Pt: 362.8 °C (760 mm Hg)
Melting Pt: 220…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06655957
CAS Number: 54232-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR