6-Bromo-5-fluoro-1H-indazole 97%

Supplier: Ambeed
Warning

A259660-10G A259660-250MG A259660-1G A259660-25G A259660-5G
103812-458EA 49 USD
103812-458 103852-340 76815-984 76815-986 77091-916 77091-918 77091-920
6-Bromo-5-fluoro-1H-indazole 97%
6-Bromo-5-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
Boiling Pt: 332.2±22.0 °C (760 mm Hg)
Flash Pt: 154.7 °C
Storage Temperature: Ambient
MDL Number: MFCD18089816
CAS Number: 1286734-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR