6-Bromo-5-chloropyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-bromo-3-chloropyridine, 6-Bromo-5-chloropyridin-3-amine
A137327-10G
A137327-250MG
A137327-1G
A137327-25G
A137327-5G
103802-442EA
7
USD
103802-442
77137-174
77137-176
77315-394
77315-396
77315-398
6-Bromo-5-chloropyridin-3-amine 97%
6-Bromo-5-chloropyridin-3-amine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol Boiling Pt: 324.9 °C (760 mm Hg) Flash Pt: 150.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09835243 CAS Number: 130284-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...