6-Bromo-4-methylquinoline 97%
Supplier: Ambeed
|
Warning
|
A104602-100MG
A104602-250MG
A104602-25G
A104602-1G
103796-740EA
48
USD
103796-740
76809-506
76809-508
76809-510
77309-322
6-Bromo-4-methylquinoline 97%
6-Bromo-4-methylquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₈BrN MW: 222.08 g/mol Boiling Pt: 315.8 °C (760 mm Hg) Melting Pt: 94…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258893 CAS Number: 41037-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...