6-Bromo-4-methylquinolin-2-ol 95%

Supplier: Ambeed
Warning

A166189-100MG A166189-250MG A166189-1G A166189-5G
103806-020EA 78 USD
103806-020 103844-280 76784-056 76784-058 77320-518 77320-520
6-Bromo-4-methylquinolin-2-ol 95%
6-Bromo-4-methylquinolin-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Boiling Pt: 392.6 °C (760 mm Hg)
Flash Pt: 191.3 °C
Storage Temperature: Ambient
MDL Number: MFCD11053820
CAS Number: 89446-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR