6-Bromo-4-methylpyridin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-bromo-4-methylpyridine

A134435-10G A134435-250MG A134435-1G A134435-25G A134435-5G A134435-100G
103802-014EA 10 USD
103802-014 76782-576 76782-578 77298-712 77314-768 77314-770 77666-946
6-Bromo-4-methylpyridin-2-amine 97%
6-Bromo-4-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.05 g/mol
Melting Pt: 115…116 °C
Flash Pt: 131.41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11036206
CAS Number: 73895-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR