6-Bromo-4-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A254748-100MG A254748-250MG
103812-162EA 99 USD
103812-162 103852-014 76771-770 76771-772
6-Bromo-4-methyl-1H-indazole 97%
6-Bromo-4-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781476
CAS Number: 885520-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR