6-Bromo-4-methyl-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A121830-250MG A121830-25G
103800-142EA 67 USD
103800-142 76766-536 76766-538
6-Bromo-4-methyl-1H-benzo[d]imidazole 97%
6-Bromo-4-methyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.06 g/mol
Boiling Pt: 420.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06411034
CAS Number: 255064-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR