6-Bromo-4-methoxy-1H-indole 95%

Supplier: Ambeed
Warning

A230869-100MG A230869-10G A230869-250MG A230869-5G A230869-1G
103810-732EA 192 USD
103810-732 103850-478 76771-142 76771-144 76771-146 77089-350 77303-890
6-Bromo-4-methoxy-1H-indole 95%
6-Bromo-4-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.07 g/mol
Boiling Pt: 346.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03095330
CAS Number: 393553-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR