6-Bromo-4-methoxy-1H-indazole 95%

Supplier: Ambeed
Warning

A197529-250MG A197529-1G
103816-930EA 49 USD
103816-930 103847-550 76770-044 77085-560
6-Bromo-4-methoxy-1H-indazole 95%
6-Bromo-4-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
Boiling Pt: 360.9±22.0 °C (760 mm Hg)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD07781361
CAS Number: 885519-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR