6-Bromo-4-iodo-1H-indazole 98%

Supplier: Ambeed
Warning

A186797-100MG A186797-1G
103815-962EA 99 USD
103815-962 103846-550 76825-112 77302-042
6-Bromo-4-iodo-1H-indazole 98%
6-Bromo-4-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MW: 322.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08689919
CAS Number: 885519-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR