6-Bromo-4-hydroxy-1H-quinolin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4-hydroxyquinolin-2(1H)-one

A219537-100MG A219537-250MG A219537-1G
103818-802EA 20 USD
103818-802 103849-678 76873-904 76873-906 77303-530
6-Bromo-4-hydroxy-1H-quinolin-2-one 97%
6-Bromo-4-hydroxy-1H-quinolin-2-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847588
CAS Number: 54675-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR