6-Bromo-4-fluoro-1H-indole 98%

Supplier: Ambeed
Warning

A343036-1G A343036-100MG A343036-10G A343036-250MG A343036-5G
103825-874EA 228 USD
103825-874 103836-848 76839-900 76989-474 76989-476 76989-478 77306-552
6-Bromo-4-fluoro-1H-indole 98%
6-Bromo-4-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781550
CAS Number: 885520-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR