6-Bromo-4-fluoro-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A237672-250MG
A237672-1G
A237672-5G
A237672-100MG
103811-170EA
27
USD
103811-170
103850-952
76771-332
77090-048
77090-050
77877-793
6-Bromo-4-fluoro-1H-indazole 98%
6-Bromo-4-fluoro-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄BrFN₂ Boiling Pt: 331.3 °C (760 mm Hg) Flash Pt: 154.15 °C Storage Temperature: Ambient |
MDL Number:
MFCD07781444 CAS Number: 885520-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...