6-Bromo-4-chlorothieno[2,3-d]pyrimidine 98%
Supplier: Ambeed
A100590-100MG
A100590-10G
A100590-250MG
A100590-25G
A100590-1G
103795-528EA
22
USD
103795-528
103821-216
76780-384
76780-386
76780-388
76780-390
77296-770
6-Bromo-4-chlorothieno[2,3-d]pyrimidine 98%
6-Bromo-4-chlorothieno[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₂BrClN₂S |
MDL Number:
MFCD09264088 CAS Number: 56844-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...