6-Bromo-4-chloroquinoline 98%
Supplier: Ambeed
|
Warning
|
A287357-1G
A287357-25G
A287357-5G
A287357-100G
A287357-10G
A287357-250MG
103813-748EA
7
USD
103813-748
103834-890
76837-434
76837-436
76837-438
77114-194
77114-196
77114-198
6-Bromo-4-chloroquinoline 98%
6-Bromo-4-chloroquinoline
Precaución: for Research Use Only
Formula:
C₉H₅BrClN MW: 242.50 g/mol Flash Pt: 144.056 °C Storage Temperature: Ambient |
MDL Number:
MFCD00511001 CAS Number: 65340-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...