6-Bromo-4-chloropicolinic acid 95%

Supplier: Ambeed
Warning

A299394-100MG A299394-250MG A299394-1G
103814-226EA 133 USD
103814-226 103835-372 77239-222 77239-224 77305-764
6-Bromo-4-chloropicolinic acid 95%
6-Bromo-4-chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188857
CAS Number: 1060811-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR