6-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A218103-100MG A218103-1MG A218103-250MG
103818-692EA 744 USD
103818-692 76986-946 76986-948 76986-950
6-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidine 95%
6-Bromo-4-chloro-5H-pyrrolo[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.46 g/mol
Boiling Pt: 384.3±37.0 °C (760 mm Hg)
Flash Pt: 186.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17012952
CAS Number: 1211536-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR