6-Bromo-4-chloro-3-nitroquinoline 97%

Supplier: Ambeed
Warning

A700952-100MG A700952-250MG
103840-640EA 184 USD
103840-640 77873-106 77873-107
6-Bromo-4-chloro-3-nitroquinoline 97%
6-Bromo-4-chloro-3-nitroquinoline

Precaución: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₉H₄BrClN₂O₂
Boiling Pt: 387.6 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09909913
CAS Number: 723281-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR