6-Bromo-4-chloro-2-methylquinazoline 98%
Supplier: Ambeed
|
Warning
|
A615143-100MG
A615143-250MG
A615143-1G
A615143-5G
103840-188EA
94
USD
103840-188
76775-334
76775-336
76844-354
76844-356
6-Bromo-4-chloro-2-methylquinazoline 98%
6-Bromo-4-chloro-2-methylquinazoline
Precaución: for Research Use Only
Formula:
C₉H₆BrClN₂ MW: 257.5143 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06657210 CAS Number: 351426-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...