6-Bromo-4-chloro-1H-indazole 96%

Supplier: Ambeed
Warning

A204036-100MG A204036-10G A204036-250MG A204036-1G A204036-5G
103817-520EA 32 USD
103817-520 103848-184 76770-248 76770-250 76770-252 77302-894 77302-896
6-Bromo-4-chloro-1H-indazole 96%

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
Melting Pt: 213…217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781350
CAS Number: 885518-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR