2-Amino-6-bromo-3-methoxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-methoxypyridin-2-amine

A484454-1G A484454-5G A484454-100MG A484454-250MG
103828-166EA 106 USD
103828-166 103839-240 76843-176 76843-178 76979-768 76979-770
2-Amino-6-bromo-3-methoxypyridine 97%
2-Amino-6-bromo-3-methoxypyridine

Precaución: For research use only

Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD15142726
CAS Number: 916737-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR